missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PBDC1 Antibody, Alexa Fluor™ 350, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-18458AF350
This item is not returnable.
View return policy
Description
PBDC1 Polyclonal antibody specifically detects PBDC1 in Human samples. It is validated for Western Blot
Specifications
| PBDC1 | |
| Polyclonal | |
| Western Blot | |
| chromosome X open reading frame 26, hypothetical protein LOC51260, MGC874,2610029G23Rik | |
| Recognizes region between residues Between Between 1 and 50.. | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| Alexa Fluor 350 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 51260 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction