missing translation for 'onlineSavingsMsg'
Learn More

PBDB-T-2F (PCE14)

Product Code. 17186534
Click to view available options
Quantity:
100 mg
250 mg
500 mg
Unit Size:
100mg
250mg
500mg
This item is not returnable. View return policy

Product Code. 17186534

Brand: Thermo Scientific Alfa Aesar H66179.MD

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 1802013-83-7 | (C{6}{8}H{7}{6}F{2}O{2}S{8})n

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1802013-83-7
Molecular Formula (C{6}{8}H{7}{6}F{2}O{2}S{8})n
Synonym PCE135

Specifications

Quantity 250 mg
Physical Form Solid
Chemical Name or Material PBDB-T-2 F (PCE14)

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.