missing translation for 'onlineSavingsMsg'
Learn More

Pazufloxacin mesylate, MedChemExpress

Product Code. 16498426
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16498426 100 mg 100mg
16432206 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16498426 Supplier Medchem Express Supplier No. HYB0724A
 Restricted Item
Estimated Shipment: 04-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Pazufloxacin (T-3761) mesylate is a fluoroquinolone antibiotic.

Pazufloxacin (T-3761) mesylate is a fluoroquinolone antibiotic.

Specifications

Chemical Name or Material Pazufloxacin mesylate
Physical Form Solid
CAS 163680-77-1
Color White
Quantity 100 mg
Purity Grade Notes Research
Molecular Formula C17H19FN2O7S
For Use With (Application) COVID-19-immunoregulation
Synonym T-3762 Pazufloxacin methanesulfonate Pazufloxacin mesilate
Solubility Information DMSO : 100 mg/mL (241.31 mM; Need ultrasonic) ∣H2O : ≥ 100 mg/mL (241.31 mM)
SMILES O=C(C(C1=O)=CN2[C@@H](C)COC3=C(C4(N)CC4)C(F)=CC1=C23)O.CS(=O)(O)=O
Molecular Weight (g/mol) 414.41
Formula Weight 414.41
Percent Purity 99.83%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.