missing translation for 'onlineSavingsMsg'
Learn More

PazePC (>90%), TRC

Product Code. 30427001
Change view
Click to view available options
Quantity:
10 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30427001 100 mg
30418858 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30427001 Supplier Toronto Research Chemicals Supplier No. TRCP210500100MG
 Restricted Item
Estimated Shipment: 18-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 117746-89-1
Molecular Formula C33H64NO10P
Molecular Weight (g/mol) 665.84
Synonym (7R)-7-[(8-Carboxy-1-oxooctyl)oxy]-4-hydroxy-N,N,N-trimethyl-10-oxo-3,5,9-trioxa-4-phosphapentacosan-1-aminium 4-Oxide Inner Salt, (R)-7-[(8-Carboxy-1-oxooctyl)oxy]-4-hydroxy-N,N,N-trimethyl-10-oxo-3,5,9-trioxa-4-phosphapentacosan-1-aminium 4-Oxide Inner Salt, 1-Palmitoyl-2-azelaoyl-sn-glycerophosphorylcholine
IUPAC Name [(2R)-2-(8-carboxyoctanoyloxy)-3-hexadecanoyloxypropyl] 2-(trimethylazaniumyl)ethyl phosphate
SMILES CCCCCCCCCCCCCCCC(=O)OC[C@H](COP(=O)([O-])OCC[N+](C)(C)C)OC(=O)CCCCCCCC(=O)O

Specifications

Quantity 100 mg
Formula Weight 665.4268
Chemical Name or Material PazePC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.