Learn More
PAXIP1 Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA553522
Description
Immunogen sequence: SNTVLICKEP TGLKYEKAKE WRIPCVNAQW LGDILLGNFE ALRQIQYSRY TAFSLQDPFA PTQHLVLNLL DAWRVPLKVS AELLMSIRLP PKLKQNEVAN VQPSSKRARI EDVPPPTKKL TPELTPFVLF TGFEPVQVQQ YIKK Highest antigen sequence identity to the following orthologs: Mouse - 88%, Rat - 90%.
This gene is a member of the paired box gene family and encodes a nuclear protein with six BRCT domains. This protein plays a critical role in maintaining genome stability, condensation of chromatin and progression through mitosis.
Specifications
| PAXIP1 | |
| Polyclonal | |
| Unconjugated | |
| PAXIP1 | |
| CAGF28, CAGF29, FLJ41049, PACIP1, PAXIP1L, PTIP, TNRC2, PAX transcription activation domain interacting protein 1 like; PAX-interacting protein 1; protein encoded by CAG trinucleotide repeats | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 22976 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Western Blot | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q6ZW49 | |
| PAXIP1 | |
| Recombit protein corresponding to Human PAXIP1 | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Research Use Only