missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Paxillin Monoclonal specifically detects Paxillin in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.
Specifications
Specifications
| Antigen | Paxillin |
| Applications | Western Blot, ELISA |
| Classification | Monoclonal |
| Clone | LD4 sAB |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Dilution | Western Blot, ELISA |
| Formulation | PBS |
| Gene Alias | FLJ16691, paxillin |
| Gene Symbols | PXN |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?