missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Paxillin Antibody (S07-1B4), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-20072-100ul
This item is not returnable.
View return policy
Description
Paxillin Monoclonal antibody specifically detects Paxillin in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| Paxillin | |
| Monoclonal | |
| 0.3 mg/mL | |
| Western Blot, Immunohistochemistry 1:20, Immunohistochemistry-Paraffin | |
| FLJ16691, paxillin | |
| A synthetic peptide of human Paxillin (Uniprot # P49023) | |
| 100 μg | |
| Cell Biology, Cellular Markers, Phospho Specific, Signal Transduction | |
| 5829 | |
| Store at 4°C short term. Store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| S07-1B4 | |
| Unconjugated | |
| 50mM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction