missing translation for 'onlineSavingsMsg'
Learn More

Paxillin Antibody (GT722), Novus Biologicals™

Product Code. 18392876 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18392876 100 μg 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18392876 Supplier Novus Biologicals Supplier No. NBP313562

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Paxillin Monoclonal antibody specifically detects Paxillin in Human, Mouse, Rat samples. It is validated for Western Blot, Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen Paxillin
Applications Western Blot, Immunofluorescence
Classification Monoclonal
Clone GT722
Conjugate Unconjugated
Dilution Western Blot, Immunofluorescence 1:100-1:1000
Formulation PBS
Gene Alias FLJ16691, paxillin
Host Species Mouse
Immunogen Carrier-protein conjugated synthetic peptide corresponding to residues around human Paxillin (non-phospho Tyr118). The exact sequence is proprietary.
Purification Method Protein G purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Cell Biology, Cellular Markers, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 5829
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.