missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PAXBP1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA552002
This item is not returnable.
View return policy
Description
Immunogen sequence: ALSSLNVLRP GEIPDAAFIH AARKKRQMAR ELGDFTPHDN EPGKGRLVRE DENDASDDED DDEKRRIVFS VKEKSQRQKI AEEIGIEGSD DDALVTGEQD EELSRWEQEQ IRKGINIPQV QASQPAEVNM YYQNT Highest antigen sequence identity to the following orthologs: Mouse - 97%, Rat - 98%.
This gene encodes a protein that may bind to GC-rich DNA sequences, which suggests its involvement in the regulation of transcription. Alternative splicing of this gene results in multiple transcript variants.
Specifications
| PAXBP1 | |
| Polyclonal | |
| Unconjugated | |
| PAXBP1 | |
| 1810007M14Rik; BM020; C21orf66; FSAP105; functional spliceosome-associated protein 105; Gcfc; GCFC1; GC-rich sequence DNA binding factor 1; GC-rich sequence DNA-binding factor 1; GC-rich sequence DNA-binding factor candidate; GC-rich sequence DNA-binding factor homolog; PAX3 and PAX7 binding protein 1; PAX3- and PAX7-binding protein 1; PAX3/7BP; Paxbp1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 94104 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunocytochemistry | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9Y5B6 | |
| PAXBP1 | |
| Recombinant protein corresponding to Human PAXBP1. Recombinant protein control fragment (Product #RP-100751). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction