missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PAX8 Mouse anti-Human, Clone: 4H7B3, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 60145-4-IG-150UL
This item is not returnable.
View return policy
Description
This gene encodes a member of the paired box family of transcription factors. Members of this gene family typically encode proteins that contain a paired box domain, an octapeptide, and a paired-type homeodomain. This nuclear protein is involved in thyroid follicular cell development and expression of thyroid-specific genes. Mutations in this gene have been associated with thyroid dysgenesis, thyroid follicular carcinomas and atypical follicular thyroid adenomas. Alternatively spliced transcript variants encoding different isoforms have been described.Specifications
| PAX8 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q06710 | |
| PAX8 | |
| PAX8 Fusion Protein Ag0306 | |
| 150 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b |
| Immunohistochemistry (Paraffin), Western Blot | |
| 4H7B3 | |
| Unconjugated | |
| PAX8 | |
| paired box 8, Paired box protein Pax 8, PAX8 | |
| Mouse | |
| Protein A | |
| RUO | |
| 7849 | |
| -20°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction