Learn More
Invitrogen™ PAX8 Monoclonal Antibody (1F8-3A8)
Mouse Monoclonal Antibody
Brand: Invitrogen™ MA1117
Description
Western blot analysis of MA1-117 detects a prominent ~48 kDa protein in human adenocarcinoma SKOV-3 and SV40 transformed African green monkey (Cercopithecus aethiops) kidney cells. In SKOV-3 cells, additional unknown low molecular bands are also detected. Specificity of the antibody was confirmed in HeLa (negative control) and 293T cells overexpressing full length PAX8.
This gene encodes a member of the paired box family of transcription factors. Members of this gene family typically encode proteins that contain a paired box domain, an octapeptide, and a paired-type homeodomain. This nuclear protein is involved in thyroid follicular cell development and expression of thyroid-specific genes. Mutations in this gene have been associated with thyroid dysgenesis, thyroid follicular carcinomas and atypical follicular thyroid adenomas. Alternatively spliced transcript variants encoding different isoforms have been described.
Specifications
| PAX8 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1 mg/mL BSA, 30% glycerol and 0.05% sodium azide | |
| Q00288, Q06710 | |
| PAX8 | |
| Purified internal fragment of human recombinant PAX8 expressed in E. coli. | |
| 100 μg | |
| Primary | |
| Human, Mouse, Monkey | |
| Antibody | |
| IgG1 |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1F8-3A8 | |
| Unconjugated | |
| PAX8 | |
| HGNC:8622; paired box 8; paired box gene 8; paired box protein Pax-8; paired domain gene 8; PAX8; Pax-8; Pax-8 protein | |
| Mouse | |
| Protein A | |
| RUO | |
| 18510, 7849 | |
| -20°C | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.