missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PAX8 Antibody (PAX8/1491), DyLight 488, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-54486G
This item is not returnable.
View return policy
Description
PAX8 Monoclonal specifically detects PAX8 in Human samples. It is validated for Western Blot, Immunohistochemistry-Paraffin.
Specifications
| PAX8 | |
| Monoclonal | |
| DyLight 488 | |
| 50 mM sodium borate with 0.05% sodium azide | |
| PAX8 | |
| Recombinant fragment (around aa 60-261) of human PAX8 protein (exact sequence is proprietary) (Uniprot: Q06710) | |
| 0.1 mL | |
| Primary | |
| Recognizes a protein of 62kDa, identified as PAX8. It is a member of the paired box (PAX) family of transcription factors. This nuclear protein is involved in thyroid follicular cell development and expression of thyroid-specific genes. Mutations in this gene have been associated with thyroid dysgenesis, thyroid follicular carcinomas, and atypical thyroid adenomas. PAX-8 is expressed in the thyroid (and associated carcinomas), non-ciliated mucosal cells of the fallopian tubes, and simple ovarian inclusion cysts, but not normal ovarian surface epithelial cells. PAX-8 is expressed in a high percentage of ovarian serous, endometrioid, and clear cell carcinomas, but only rarely in primary ovarian mucinous adenocarcinomas. PAX-8 expression is reported in renal tubules as well as renal cell carcinoma, nephroblastoma, and seminoma. PAX-8 antibody may be used as an additional immunohistochemical marker for renal epithelial tumors. | |
| Store at 4°C in the dark. |
| Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| PAX8/1491 | |
| Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin | |
| paired box 8, paired box gene 8, paired box protein Pax-8, paired domain gene 8 | |
| Mouse | |
| Protein A or G purified | |
| Cancer | |
| 7849.0 | |
| Human | |
| IgG2b κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction