missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PAX7 Mouse, Clone: PAX7/497, enQuireBio™
Mouse Monoclonal Antibody
Brand: EnquireBio 5081-MSM1-P0
This item is not returnable.
View return policy
Specifications
| PAX7 | |
| Nuclear | |
| PAX7/497 | |
| Unconjugated | |
| NP_039236 | |
| Mouse | |
| 57kDa | |
| Protein A/G Purified | |
| RUO | |
| 5081 | |
| IgG1 κ |
| Western Blot | |
| Monoclonal | |
| 200μg/mL | |
| 10mM PBS with 0.05% BSA and 0.05% Azide | |
| PAX7 | |
| Recombinant fragment (aa301-505) of human PAX7 protein | |
| HeLa, Jurkat, A431, MCF-7, U2OS or NIH/3T3 cells. Rhabdomyosarcoma (RMS) | |
| 20 μg | |
| Primary | |
| Short term: Store at 2-4°C. Long term, aliquot and store at -20°C. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction