missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pax6 Rabbit anti-Human, Clone: PAX6/7078R, Novus Biologicals™
Rabbit Monoclonal Antibody
£228.00 - £411.00
Specifications
| Antigen | Pax6 |
|---|---|
| Clone | PAX6/7078R |
| Concentration | 0.2 mg/ml |
| Dilution | Western Blot 1-2 ug/ml, Flow Cytometry 0.5-1 ug/million cells, Immunofluorescence 0.5-1 ug/ml, Immunohistochemistry-Paraffin |
| Applications | Western Blot, Flow Cytometry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18306492
|
Novus Biologicals
NBP3-13988-20UG |
20 μg |
£228.00
20µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18350874
|
Novus Biologicals
NBP3-13988-100UG |
100 μg |
£411.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Pax6 Monoclonal antibody specifically detects Pax6 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunofluorescence, Immunohistochemistry (Paraffin)Specifications
| Pax6 | |
| 0.2 mg/ml | |
| Western Blot, Flow Cytometry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Cellular Markers, Diabetes Research, Neuroscience, Sensory Systems, Stem Cell Markers, Stem Cells, Transcription Factors and Regulators, Vision | |
| 10 mM PBS with 0.05% BSA | |
| keratitis), MGC17209, Oculorhombin, paired box 6, paired box protein Pax-6 | |
| Recombinant fragment (N-terminus; aa 1-300) of human Pax6 protein (Uniprot: P26367) | |
| Primary | |
| Store at 4°C. |
| PAX6/7078R | |
| Western Blot 1-2 ug/ml, Flow Cytometry 0.5-1 ug/million cells, Immunofluorescence 0.5-1 ug/ml, Immunohistochemistry-Paraffin | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| P26367 | |
| 5080 | |
| IgG κ | |
| Protein A or G purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title