missing translation for 'onlineSavingsMsg'
Learn More

Pax6 Antibody (PAX6/7078R), Janelia Fluor™ 549, Novus Biologicals™

Product Code. 18325154 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18325154 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18325154 Supplier Novus Biologicals Supplier No. NBP314033JF549

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

Pax6 Monoclonal antibody specifically detects Pax6 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen Pax6
Applications Western Blot, Flow Cytometry, Immunofluorescence
Classification Monoclonal
Clone PAX6/7078R
Conjugate Janelia Fluor 549
Formulation 50mM Sodium Borate
Gene Alias keratitis), MGC17209, Oculorhombin, paired box 6, paired box protein Pax-6
Host Species Rabbit
Immunogen Recombinant fragment (N-terminus; aa 1-300) of human Pax6 protein (Uniprot: P26367)
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cellular Markers, Diabetes Research, Neuroscience, Sensory Systems, Stem Cell Markers, Stem Cells, Transcription Factors and Regulators, Vision
Primary or Secondary Primary
Gene ID (Entrez) 5080
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.