missing translation for 'onlineSavingsMsg'
Learn More

Pax6 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 3 publications

Brand:  Novus Biologicals NB300-750-0.025mg

 View more versions of this product

Product Code. 18540533

  • £174.00 / 25µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Pax6 Polyclonal specifically detects Pax6 in Human, Mouse, Rat, Fish, Primate, Monkey, Sheep samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Pax6
Polyclonal
Unconjugated
PBS, 1mg/ml BSA with 0.05% Sodium Azide
keratitis), MGC17209, Oculorhombin, paired box 6, paired box protein Pax-6
Rabbit
47 kDa
0.025 mg
Cellular Markers, Diabetes Research, Neuronal Stem Cell Markers, Neuronal Stem Cells, Neuroscience, Sensory Systems, Stem Cell Markers, Stem Cells, Transcription Factors and Regulators, Vision
5080
Human
IgG
Western Blot
1 mg/mL
Western Blot 1:1000, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1:50, Immunoprecipitation 5 μg, Immunocytochemistry 1:100
P63016
PAX6
Synthetic peptide corresponding to residues C R(267) E E K L R N Q R R Q A S N T P S H I(285) of mouse PAX6.
Affinity Purified
RUO
Primary
Detects a ∼47 kDa protein representing PAX6 (Schwann Cell Marker) from rat whole eye extracts and ovine retinal extracts.
Store at -20°C. Avoid freeze/thaw cycles.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only