missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pax6 Antibody (CL5414) [Alexa Fluor™ 594], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-61411AF594
This item is not returnable.
View return policy
Description
Pax6 Monoclonal antibody specifically detects Pax6 in Human,Mouse,Rat samples. It is validated for Immunohistochemistry,Immunohistochemistry (Paraffin),Immunocytochemistry/ Immunofluorescence
Specifications
| Pax6 | |
| Monoclonal | |
| Alexa Fluor 594 | |
| keratitis), MGC17209, Oculorhombin, paired box 6, paired box protein Pax-6 | |
| This antibody was developed using a recombinant protein derived from P26367, with the exact immunogen sequence remaining proprietary. | |
| 0.1 mL | |
| Cellular Markers, Diabetes Research, Neuroscience, Sensory Systems, Stem Cell Markers, Stem Cells, Transcription Factors and Regulators, Vision | |
| 5080 | |
| Store at 4⪚C in the dark. | |
| IgG1 |
| Immunohistochemistry, Immunohistochemistry (Paraffin), Immunocytochemistry/Immunofluorescence | |
| CL5414 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction