Learn More
PAX5 Monoclonal Antibody (1H9), eBioscience™, Invitrogen™
Rat Monoclonal Antibody
£146.00 - £362.00
Specifications
Clone | 1H9 |
---|---|
Concentration | 0.5 mg/mL |
Classification | Monoclonal |
Conjugate | Unconjugated |
Form | Liquid |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15278237
|
Affymetrix eBioscience
14-9918-82 |
100 μg |
£362.00
100µg |
Estimated Shipment: 05-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15268237
|
Affymetrix eBioscience
14-9918-80 |
25 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
Description: The monoclonal antibody 1H9 recognizes both mouse and human Pax5. Pax5, also known as BSAP (B cell specific activator protein), is a member of the paired box (pax) family of transcription factors. Pax5 is the only member of the pax family of transcription factors that is expressed in hematopoietic cells. During embryogenesis, Pax5 is transiently expressed in the brain of mice and in the mesencephalon and spinal cord of humans. Its expression is upregulated early in B cell development at the time of B cell commitment and is maintained throughout most subsequent stages. Suppression of Pax5 is essential for expression of Blimp-1 and the terminal differentiation of plasma cells. In the spleen, expression of Pax5 is higher in marginal zone B cells (B220+ CD21high CD23low) than in other B cells, especially the transition 1 stage (B220+ CD21- CD23-). In addition to its role in B cell development, Pax5 also affects VH-DJH heavy chain recombination as well as influencing the expression of many other B and non-B cell related proteins. Pax5 expression is correlated with many neoplasms. In diffuse large B cell lymphomas (DLBCL) and non-Hodgkin lymphomas Pax5 is often mutated while in B-cell ALL, expression levels are high. Additionally, translocation with elastin, IGH, ETV6, FOXP1, and EVI3 have been identified. eBioscience recommends that the Foxp3 Buffers (cat.
PAX5 is a member of the paired box (PAX) family of transcription factors. The PAX proteins are important regulators in early development and alterations in the expression of their genes are thought to contribute to neoplastic transformation. PAX5 is the B-cell lineage specific activator protein (BSAP) that is expressed at early, but not late stages, of B-cell differentiation. Its expression has also been detected in developing CNS and testis, therefore, PAX5 may not only play an important role in B-cell differentiation, but also in neural development and spermatogenesis. Mutations in the gene can result in leukemia, acute lymphoblastic 3.Specifications
1H9 | |
Monoclonal | |
Liquid | |
RUO | |
Q02548, Q02650 | |
PAX5 | |
Primary | |
4° C | |
PAX5 |
0.5 mg/mL | |
Unconjugated | |
Rat | |
Human, Mouse | |
18507, 5079 | |
IgG2a κ | |
Affinity chromatography | |
Antibody |
For Research Use Only.