missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pax5/BSAP Mouse anti-Human, Clone: PAX5/3735, Novus Biologicals™
Mouse Monoclonal Antibody
£396.00
Specifications
| Antigen | Pax5/BSAP |
|---|---|
| Clone | PAX5/3735 |
| Concentration | 0.2 mg/mL |
| Dilution | Western Blot, Immunohistochemistry-Paraffin 1 to 2 μg/mL, Protein Array |
| Applications | Western Blot, Immunohistochemistry (Paraffin), Peptide Array |
Description
Pax5/BSAP Monoclonal specifically detects Pax5/BSAP in Human samples. It is validated for Western Blot, Immunohistochemistry-Paraffin, Protein Array.Specifications
| Pax5/BSAP | |
| 0.2 mg/mL | |
| Western Blot, Immunohistochemistry (Paraffin), Peptide Array | |
| Unconjugated | |
| Mouse | |
| B Cell Development and Differentiation Markers, Cancer, Transcription Factors and Regulators | |
| 10 mM PBS with 0.05% BSA | |
| 5079 | |
| IgG1 λ | |
| Protein A or G purified |
| PAX5/3735 | |
| Western Blot, Immunohistochemistry-Paraffin 1 to 2 μg/mL, Protein Array | |
| Monoclonal | |
| Purified | |
| RUO | |
| Canine, Equine, Feline, Human, Rhesus Macaque | |
| B cell specific activator protein, B-cell-specific transcription factor, BSAPB-cell lineage specific activator, paired box 5, paired box gene 5 (B-cell lineage specific activator protein), paired box gene 5 (B-cell lineage specific activator), paired box homeotic gene 5, paired box protein Pax-5, transcription factor PAX 5 | |
| A recombinant fragment (around aa 235-382) of human Pax5/BSAP protein (exact sequence is proprietary) | |
| Primary | |
| Store at 4°C. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title