Learn More
Pax5/BSAP Antibody (rPAX5/4228), HRP, Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP3-08419H
Product Code. 18640029
  • £471.00 / 100µL
Description
Pax5/BSAP Monoclonal antibody specifically detects Pax5/BSAP in Human, Canine samples. It is validated for Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Pax5/BSAP
Monoclonal
HRP
B cell specific activator protein, B-cell-specific transcription factor, BSAPB-cell lineage specific activator, paired box 5, paired box gene 5 (B-cell lineage specific activator protein), paired box gene 5 (B-cell lineage specific activator), paired box homeotic gene 5, paired box protein Pax-5, transcription factor PAX 5
A recombinant fragment (around aa 235-382) of human PAX5 protein (exact sequence is proprietary) (Uniprot: Q02548)
100 μg
B Cell Development and Differentiation Markers, Cancer, Transcription Factors and Regulators
5079
Store at 4°C in the dark.
IgG2a κ
Immunohistochemistry (Paraffin)
rPAX5/4228
PBS
Mouse
Protein A or G purified
RUO
Primary
Human, Canine
Purified