missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Pax5/BSAP Antibody (CL14548), Novus Biologicals™

Mouse Monoclonal Antibody

£298.00 - £449.00

Specifications

Antigen Pax5/BSAP
Clone CL14548
Concentration 1 mg/mL
Dilution Western Blot 1 μg/mL, Immunohistochemistry 1:50-1:200
Applications Immunohistochemistry, Western Blot
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
30043505
View Documents
Novus Biologicals™
NBP3-26864-25ul
25 μL
£298.00
25µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
30043958
View Documents
Novus Biologicals™
NBP3-26864-100ul
100 μL
£449.00
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Pax5/BSAP Monoclonal antibody specifically detects Pax5/BSAP in Human samples. It is validated for Immunohistochemistry,Western Blot
Specifications

Specifications

Pax5/BSAP
1 mg/mL
Immunohistochemistry, Western Blot
Unconjugated
Mouse
B Cell Development and Differentiation Markers, Cancer, Transcription Factors and Regulators
PBS, pH 7.2, 40% glycerol
5079
IgG2b
Protein A purified
CL14548
Western Blot 1 μg/mL, Immunohistochemistry 1:50-1:200
Monoclonal
Purified
RUO
Human
B cell specific activator protein, B-cell-specific transcription factor, BSAPB-cell lineage specific activator, paired box 5, paired box gene 5 (B-cell lineage specific activator protein), paired box gene 5 (B-cell lineage specific activator), paired box homeotic gene 5, paired box protein Pax-5, transcription factor PAX 5
This antibody was developed against a Synthetic Peptide of Q02548. Actual immunogen sequence is proprietary.
Primary
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.