missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PAX3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 21386-1-AP-20UL
This item is not returnable.
View return policy
Description
Pax 3 (Paired Box 3) is a transcription factor that is involved in development of the peripheral nervous system, melanocytes, some vascular smooth muscle, and a number of other derivatives. It regulates neurogenesis in pre-migratory neural crest cells from the dorsal neural tube, and in myogenic progenitors in the presomitic mesoderm and the hypaxial somites. Alternative splicing results in isoforms having different C-termini.Specifications
| PAX3 | |
| Polyclonal | |
| Unconjugated | |
| PAX3 | |
| CDHS, HUP2, paired box 3, Paired box protein Pax 3, PAX3, WS1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 114502, 18505, 5077 | |
| -20°C | |
| Liquid |
| Western Blot, Immunoprecipitation | |
| 0.59 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P23760, P24610 | |
| PAX3 | |
| PAX3 Fusion Protein Ag16021 | |
| 20 μL | |
| Primary | |
| Human, Rat, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction