missing translation for 'onlineSavingsMsg'
Learn More

Pax2 Antibody (PAX2/8671R), Novus Biologicals™

Rabbit Monoclonal Antibody

£224.00 - £412.00

Specifications

Antigen Pax2
Clone PAX2/8671R
Concentration 0.2 mg/ml
Dilution Immunohistochemistry-Paraffin 1-2 μg/mL
Applications Immunohistochemistry (Paraffin)
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18604924
View Documents
Novus Biologicals
NBP3-20575-20ug
20 μg
£224.00
20µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18690444
View Documents
Novus Biologicals
NBP3-20575-100ug
100 μg
£412.00
100µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Pax2 Monoclonal antibody specifically detects Pax2 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications

Specifications

Pax2
0.2 mg/ml
Immunohistochemistry (Paraffin)
Unconjugated
Rabbit
Chaperone Mediated Autophagy (CMA), Neuronal Cell Markers, Stem Cell Markers
10mM PBS with 0.05% BSA
5076
IgG κ
Protein A or G purified
PAX2/8671R
Immunohistochemistry-Paraffin 1-2 μg/mL
Monoclonal
Purified
RUO
Human
paired box 2, paired box gene 2, paired box homeotic gene 2, paired box protein Pax-2
Recombinant fragment (around aa 223-354) of human Pax2 protein (exact sequence is proprietary)
Primary
Store at 4&drg;C. Do not freeze.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.