missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pax2 Antibody (CL9469) [DyLight 350], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-88934UV
This item is not returnable.
View return policy
Description
Pax2 Monoclonal antibody specifically detects Pax2 in Human samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin),Immunocytochemistry/ Immunofluorescence
Specifications
| Pax2 | |
| Monoclonal | |
| DyLight 350 | |
| paired box 2, paired box gene 2, paired box homeotic gene 2, paired box protein Pax-2 | |
| This antibody was developed using a synthetic peptide derived from Q02962, with the exact immunogen sequence remaining proprietary. | |
| 0.1 mL | |
| Chaperone Mediated Autophagy (CMA), Neuronal Cell Markers, Stem Cell Markers | |
| 5076 | |
| Store at 4⪚C in the dark. | |
| IgG1 |
| Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry/Immunofluorescence | |
| CL9469 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction