missing translation for 'onlineSavingsMsg'
Learn More

Pax2 Antibody, Alexa Fluor™ 350, Novus Biologicals™

Product Code. 18320717 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18320717 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18320717 Supplier Novus Biologicals Supplier No. NBP299013AF350

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Pax2 Polyclonal antibody specifically detects Pax2 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin).
TRUSTED_SUSTAINABILITY

Specifications

Antigen Pax2
Applications Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Alexa Fluor 350
Formulation 50 mM Sodium Borate
Gene Alias paired box 2, paired box gene 2, paired box homeotic gene 2, paired box protein Pax-2
Host Species Rabbit
Immunogen Produced in rabbits immunized with a synthetic peptide corresponding to the center region of human Pax2.
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Chaperone Mediated Autophagy (CMA), Neuronal Cell Markers, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 5076
Target Species Human, Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.