missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PAX1 Polyclonal Antibody

Description
Immunogen sequence: GTAYSPKMED WAGVNR.
The PAX genes, including PAX1, are a highly conserved family of developmental control genes that encode transcription factors and have been shown to play a role in pattern formation during embryogenesis in vertebrates. See PAX7 (MIM 167410) for a discussion of paired box domain genes.
Specifications
Specifications
| Antigen | PAX1 |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Concentration | 0.73 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.05% ProClin 300; pH 7.3 |
| Gene | PAX1 |
| Gene Accession No. | P15863 |
| Gene Alias | hbs; hunchback; HuP48; OFC2; paired box 1; paired box gene 1; Paired box homeotic gene 1; paired box protein Pax-1; paired domain gene HuP48; PAX1; Pax-1; un; undulated; wavy tail; wt |
| Gene Symbols | PAX1 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?