missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PAX1 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
£151.00 - £366.00
Specifications
| Antigen | PAX1 |
|---|---|
| Dilution | Western Blot 1:500-1:2000 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18689930
|
Novus Biologicals
NBP2-93575-0.02ml |
0.02 mL |
£151.00
0.02mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18662431
|
Novus Biologicals
NBP2-93575-0.1ml |
0.1 mL |
£366.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
PAX1 Polyclonal antibody specifically detects PAX1 in Human samples. It is validated for Western BlotSpecifications
| PAX1 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human | |
| paired box 1, paired domain gene HuP48 | |
| A synthetic peptide corresponding to a sequence within amino acids 300-400 of human PAX1 (NP_006183.2). GTAYSPKMEDWAGVNR | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
| Western Blot 1:500-1:2000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| PBS (pH 7.3), 50% glycerol | |
| 5075 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title