missing translation for 'onlineSavingsMsg'
Learn More

PAWR/ PAR4 Antibody, mFluor Violet 610 SE, Novus Biologicals™

Rabbit Polyclonal Antibody

Brand:  Novus Biologicals NB600-219MFV610

Product Code. 18462313

  • £411.00 / 0.10mL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

PAWR / PAR4 Polyclonal antibody specifically detects PAWR / PAR4 in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY
Specifications

Specifications

PAWR / PAR4
Polyclonal
Western Blot
Par-4, PAR4prostate apoptosis response protein 4, PRKC apoptosis WT1 regulator protein, PRKC, apoptosis, WT1, regulator, Prostate apoptosis response 4 protein, prostate apoptosis response protein PAR-4, transcriptional repressor PAR4, WT1-interacting protein
Synthetic peptides corresponding to the N-terminal sequence (MATGGYRTSSGLGGS) and C-terminal sequence (QENKTLLKVVGQLTR) of human PAR4.
0.1 mL
Apoptosis, Cancer, GPCR, Signal Transduction, Transcription Factors and Regulators, Tumor Suppressors
5074
Store at 4C in the dark.
IgG
Western Blot
mFluor Violet 610 SE
50mM Sodium Borate
Rabbit
Immunogen affinity purified
RUO
Primary
Human
Purified
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.