missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PAWR/ PAR4 Antibody (3G9H7), Novus Biologicals™
Mouse Monoclonal Antibody
£196.00
Specifications
| Antigen | PAWR / PAR4 |
|---|---|
| Clone | 3G9H7 |
| Dilution | Western Blot 1:500-1:2000, ELISA 1:10000, Immunohistochemistry 1:200-1:1000, Immunohistochemistry-Paraffin 1:200-1:1000 |
| Applications | Western Blot |
| Classification | Monoclonal |
Description
PAWR / PAR4 Monoclonal specifically detects PAWR / PAR4 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| PAWR / PAR4 | |
| Western Blot 1:500-1:2000, ELISA 1:10000, Immunohistochemistry 1:200-1:1000, Immunohistochemistry-Paraffin 1:200-1:1000 | |
| Monoclonal | |
| Ascites | |
| RUO | |
| Human | |
| Par-4, PAR4prostate apoptosis response protein 4, PRKC apoptosis WT1 regulator protein, PRKC, apoptosis, WT1, regulator, Prostate apoptosis response 4 protein, prostate apoptosis response protein PAR-4, transcriptional repressor PAR4, WT1-interacting protein | |
| PAWR | |
| IgG1 | |
| Unpurified | |
| Specific for PAWR PAR4. |
| 3G9H7 | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| Apoptosis, Cancer, GPCR, Signal Transduction, Transcription Factors and Regulators, Tumor Suppressors | |
| Ascites with 0.03% Sodium Azide | |
| 5074 | |
| Purified recombinant fragment of PAWR PAR4 (3G9H7) expressed in E. Coli. | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title