missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PATZ1 Recombinant Rabbit Monoclonal Antibody (6H7A4)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA557876
This item is not returnable.
View return policy
Description
Immunogen sequence: PFPSVASSAP PLTGKRGRGR PRKANLLDSM FGSPGGLREA GILPCGLCGK VFTDANRLRQ HEAQHGVTSL QLGYIDLPPP RLGENGLPIS EDPDGPRKRS R.
The protein encoded by this gene contains an A-T hook DNA binding motif which usually binds to other DNA binding structures to play an important role in chromatin modeling and transcription regulation. Its Poz domain is thought to function as a site for protein-protein interaction and is required for transcriptional repression, and the zinc-fingers comprise the DNA binding domain. Since the encoded protein has typical features of a transcription factor, it is postulated to be a repressor of gene expression. In small round cell sarcoma, this gene is fused to EWS by a small inversion of 22q, then the hybrid is thought to be translocated (t(1;22)(p36. 1;q12). The rearrangement of chromosome 22 involves intron 8 of EWS and exon 1 of this gene creating a chimeric sequence containing the transactivation domain of EWS fused to zinc finger domain of this protein. This is a distinct example of an intra-chromosomal rearrangement of chromosome 22. Four alternatively spliced transcript variants are described for this gene.
Specifications
| PATZ1 | |
| Recombinant Monoclonal | |
| 1.02 mg/mL | |
| PBS with 0.05% BSA, 50% glycerol and 0.05% ProClin 300; pH 7.3 | |
| Q9HBE1 | |
| PATZ1 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 250-350 of human PATZ1 (NP_055138.2). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| ELISA, Western Blot | |
| 6H7A4 | |
| Unconjugated | |
| PATZ1 | |
| 8430401L15Rik; AI447519; BTB/POZ domain zinc finger transcription factor; BTB-POZ domain zinc finger transcription factor; dJ400N23; Mazr; MAZ-related factor; Patz; PATZ1; POZ (BTB) and AT hook containing zinc finger 1; POZ-, AT hook-, and zinc finger-containing protein 1; POZ/BTB and AT hook containing zinc finger 1; POZ-AT hook-zinc finger protein; protein kinase A RI subunit alpha-associated protein; RIAZ; transcription factor (MAZ-related) MAZR; ZBTB19; Zfp278; zi; Zinc finger and BTB domain-containing protein 19; zinc finger protein 278; Zinc finger sarcoma gene protein; ZNF278; ZSG | |
| Rabbit | |
| Protein A | |
| RUO | |
| 23598 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction