missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PATZ1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA561616
This item is not returnable.
View return policy
Description
Immunogen sequence: LLDSMFGSPG GLREAGILPC GLCGKVFTDA NRLRQHEAQH GVTSLQLGYI DLPPPRLGEN GLPISEDPDG PRKRSRTRKQ VACEI Highest antigen sequence identity to the following orthologs: Mouse - 100%, Rat - 100%.
The protein encoded by this gene contains an A-T hook DNA binding motif which usually binds to other DNA binding structures to play an important role in chromatin modeling and transcription regulation. Its Poz domain is thought to function as a site for protein-protein interaction and is required for transcriptional repression, and the zinc-fingers comprise the DNA binding domain. Since the encoded protein has typical features of a transcription factor, it is postulated to be a repressor of gene expression. In small round cell sarcoma, this gene is fused to EWS by a small inversion of 22q, then the hybrid is thought to be translocated (t(1;22)(p36. 1;q12). The rearrangement of chromosome 22 involves intron 8 of EWS and exon 1 of this gene creating a chimeric sequence containing the transactivation domain of EWS fused to zinc finger domain of this protein. This is a distinct example of an intra-chromosomal rearrangement of chromosome 22. Four alternatively spliced transcript variants are described for this gene.
Specifications
| PATZ1 | |
| Polyclonal | |
| Unconjugated | |
| PATZ1 | |
| 8430401L15Rik; AI447519; BTB/POZ domain zinc finger transcription factor; BTB-POZ domain zinc finger transcription factor; dJ400N23; Mazr; MAZ-related factor; Patz; PATZ1; POZ (BTB) and AT hook containing zinc finger 1; POZ-, AT hook-, and zinc finger-containing protein 1; POZ/BTB and AT hook containing zinc finger 1; POZ-AT hook-zinc finger protein; protein kinase A RI subunit alpha-associated protein; RIAZ; transcription factor (MAZ-related) MAZR; ZBTB19; Zfp278; zi; Zinc finger and BTB domain-containing protein 19; zinc finger protein 278; Zinc finger sarcoma gene protein; ZNF278; ZSG | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 23598 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| ChIP Assay, Immunohistochemistry (Paraffin), Immunocytochemistry | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9HBE1 | |
| PATZ1 | |
| Recombinant protein corresponding to Human PATZ1. Recombinant protein control fragment (Product #RP-100846). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction