missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PATZ Antibody (OTI8D6), DyLight 550, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-73256R
This item is not returnable.
View return policy
Description
PATZ Monoclonal specifically detects PATZ in Human, Mouse, Rat samples. It is validated for Western Blot.
Specifications
| PATZ | |
| Monoclonal | |
| DyLight 550 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| PATZ1 | |
| Full length recombinant protein of human PATZ (NP_114440) produced in E.coli. | |
| 0.1 mL | |
| 23598.0 | |
| Store at 4°C in the dark. |
| Western Blot | |
| OTI8D6 | |
| Western Blot | |
| dJ400N23, MAZRPOZ-, AT hook-, and zinc finger-containing protein 1, PATZZinc finger and BTB domain-containing protein 19, POZ (BTB) and AT hook containing zinc finger 1, Protein kinase A RI subunit alpha-associated protein, RIAZBTB/POZ domain zinc finger transcription factor, ZBTB19BTB-POZ domain zinc finger transcription factor, Zinc finger protein 278Zinc finger sarcoma gene protein, ZNF278POZ-AT hook-zinc finger protein, ZSGMAZ-related factor | |
| Mouse | |
| Affinity Purified | |
| Primary | |
| Human, Mouse, Rat | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction