missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PATZ Antibody, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-33488-100ul
This item is not returnable.
View return policy
Description
PATZ Monoclonal antibody specifically detects PATZ in Human samples. It is validated for ELISA,Western Blot
Specifications
| PATZ | |
| Monoclonal | |
| Western Blot 1:500 - 1:1000, ELISA Recommended starting concentration is 1 μg/mL | |
| dJ400N23, MAZRPOZ-, AT hook-, and zinc finger-containing protein 1, PATZZinc finger and BTB domain-containing protein 19, POZ (BTB) and AT hook containing zinc finger 1, Protein kinase A RI subunit alpha-associated protein, RIAZBTB/POZ domain zinc finger transcription factor, ZBTB19BTB-POZ domain zinc finger transcription factor, Zinc finger protein 278Zinc finger sarcoma gene protein, ZNF278POZ-AT hook-zinc finger protein, ZSGMAZ-related factor | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 250-350 of human PATZ (NP_055138.2).,, Sequence:, PFPSVASSAPPLTGKRGRGRPRKANLLDSMFGSPGGLREAGILPCGLCGKVFTDANRLRQHEAQHGVTSLQLGYIDLPPPRLGENGLPISEDPDGPRKRSR | |
| 100 μL | |
| Primary | |
| Human | |
| Purified |
| ELISA, Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol, 0.05% BSA | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 23598 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction