missing translation for 'onlineSavingsMsg'
Learn More

Patulin, TRC

Product Code. 30040581
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
Unit Size:
10mg
25mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30040581 5 mg 5mg
30007330 10 mg 10mg
30006692 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30040581 Supplier Toronto Research Chemicals Supplier No. TRCP2065005MG
 Restricted Item
Estimated Shipment: 08-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 149-29-1
Molecular Formula C7 H6 O4
Molecular Weight (g/mol) 154.12
Synonym Patulin (6CI), 4-Hydroxy-4H-furo[3,2-c]pyran-2(6H)-one, Acetic acid, (2,4-dihydroxy-2H-pyran-3(6H)-ylidene)-, 3,4-lactone, Clairformin, Clavacin, Clavatin, Claviformin, DL-Patulin, Expansin, Expansin (antibiotic), Expansine, Gigantin, Leucopin, Mycoin, Mycoin C, Mycoin C3, NSC 32951, NSC 8120, Patuline, Penicidin, Terinin
IUPAC Name 4-hydroxy-4,6-dihydrofuro[3,2-c]pyran-2-one
SMILES OC1OCC=C2OC(=O)C=C12

Specifications

Quantity 5 mg
InChI Formula InChI=1S/C7H6O4/c8-6-3-4-5(11-6)1-2-10-7(4)9/h1,3,7,9H,2H2
Formula Weight 154.0266
Percent Purity >95%
Chemical Name or Material Patulin
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.