missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DiscoverX PathHunter™ U2OS RANK NF-kB Signaling Pathway Reporter Cell Line
Click to view available options
Quantity:
2 Vials
Unit Size:
2 ampoules
Description
The PathHunter™ U2OS RANK NF-kB Pathway Reporter Cell Line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) affecting the RANK NF-kB pathway. This cell line enables assessment of RANK NF-kB pathway stimulation based on changes in the expression of a RANK NF-kB response element-driven reporter protein. This product uses EFC technology. The included cell line expresses PL-tagged reporter protein under the control of a RANK NF-kB pathway-inducible transcriptional response element. Pathway activation induces expression of the PL-tagged protein, allowing detection of EFC signal upon addition of EA-containing detection reagent. This stable modified cell line is provided as two vials of cryopreserved cells.
Specifications
Specifications
| Assay Entry | Transcriptional Reporter |
| Cell Line | U2OS |
| For Use With (Application) | Cell Lines |
| Primary Agonist | soluble RANKL |
| Quantity | 2 Vials |
| Accession Number | NM_003839.3 |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction