missing translation for 'onlineSavingsMsg'
Learn More

DiscoverX PathHunter™ U2OS ErbB2/ErbB3 Dimerization Cell Line

Product Code. 17304289
Encompass
Click to view available options
Quantity:
2 Vials
Unit Size:
2 ampoules
This item is not returnable. View return policy

Product Code. 17304289

Brand: DiscoverX 931042C3

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

The PathHunter™ U2OS ErbB2/ErbB3 Dimerization Cell Line measures ligand-induced dimerization of ErbB2/ErbB3 . This cell line ships as two vials of cryopreserved cells.

The PathHunter™ ErbB2/ErbB3 Dimerization cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting receptor-receptor interaction. This cell line enables assessment of ligand (e.g. neuregulin-1 Beta 2) induced dimerization of ErbB2/ErbB3 receptors. This product uses EFC technology. The cell line included overexpresses ErbB2 target protein fused to PK alongside ErbB3 target protein fused to EA. Upon ligand-induced dimerization of ErbB2/ErbB3, detectable EFC signal is produced. This stable modified cell line is provided as two vials of cryopreserved cells.

Specifications

Assay Entry Dimerization
Cell Line U2OS
For Use With (Application) Cell Lines
Primary Agonist Recombinant Human Neuregulin-1 Beta 2
Quantity 2 Vials
Target Entry ErbB2/ErbB3
Accession Number NM_004448.2
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.