missing translation for 'onlineSavingsMsg'
Learn More

DiscoverX PathHunter™ HEK 293 Tie2 Functional Assay

Product Code. 17384249
Encompass
Change view
Click to view available options
Quantity:
2 Vials
Unit Size:
2 ampoules
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17384249 2 Vials 2 ampoules
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17384249 Supplier DiscoverX Supplier No. 931024C1

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

The PathHunter™ HEK 293 Tie2 Functional Assay measures Tie2 receptor tyrosine kinase activity via ligand-induced SH2 domain protein recruitment. This cell line ships as two vials of cryopreserved cells.

The PathHunter™ HEK 293 Tie2 Functional Assay provides an easy-to-use cell based assay to study drug candidates (small molecules or biologics) targeting RTKs. This kit assesses RTK activation via detection of activated Tie2 dimerization-induced SH2 domain protein recruitment. This product uses EFC technology. The included cell line overexpresses PK-tagged Tie2 and an EA-tagged SH2 domain protein. Upon ligand-induced receptor activation, SH2-EA recruitment leads to production of EFC signal. This stable modified cell line is provided as two vials of cryopreserved cells.

Specifications

Assay Entry SH2 Recruitment
Cell Line 293H
For Use With (Application) Cell Lines
Primary Agonist Angiopoietin-2
Quantity 2 Vials
Target Entry Tie2
Accession Number NM_000459.4
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.