Learn More
DiscoverX PathHunter™ β-Arrestin2 Retroparticles
Product Code. 17334369
Encompass
Change view
Click to view available options
Quantity:
4 Vials x 0.5 mL
Unit Size:
4 ampoules
Product Code. Quantity Unit Size
17334369 4 Vials x 0.5 mL 4 ampoules
1 options
Product Code. 17334369 Supplier DiscoverX Supplier No. 931087
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
The PathHunter™ β-Arrestin2 Retroparticles are retroparticles for transducing cells with β-Arrestin2-EA to allow for arrestin recruitment assays. Contains 4 vials of cryopreserved retroparticles.
The PathHunter™ β-Arrestin2 Retroparticles are MMLV particles designed to transduce cells of interest with β-Arrestin2 fused to EA. In conjunction with a plasmid expressing a PK-tagged GPCR of interest, this allows for these cells to be used for β-Arrestin2 recruitment assays. This product uses EFC technology. Ligand induced recruitment of β-Arrestin2 to the activated GPCR leads to complementation and produces EFC signal. These retroparticles ship as 4 vials of cryopreserved retroparticles.
Specifications
For Use With (Application) Retroparticles
Product Type Retroparticles
Quantity 4 Vials x 0.5 mL