missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ Path-ID™ qPCR Master Mix with Manual

Product Code. p-7181611 Shop All Applied Biosystems Products
Click to view available options
No. of Reactions:
100 Reactions
500 Reactions
Unit Size:
100 reactions
500 reactions
This item is not returnable. View return policy

Product Code. 30057788

Brand: Applied Biosystems™ 4388643M

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Ambion™ Path-ID™ qPCR Master Mix is a convenient enzyme, buffer, and dNTP mix for TaqMan™ probe-based, real-time PCR detection of DNA targets.

Ambion™ Path-ID™ qPCR Master Mix is optimized for targets of importance in animal health and to function in the presence of PCR inhibitors. The mix is supplied in one tube suitable for 100 reactions. With the exception of PCR primers and probes for your target, the master mix contains everything you need for real-time PCR, including:

  • AmpliTaq Gold™ ultrapure thermostable DNA polymerase designed for automatic hot start, to minimize nonspecific PCR products
  • Optimized buffer and dNTP concentration, for reliable PCR
  • Passive internal reference, based on proprietary ROX™ dye, for precise data analysis.
TRUSTED_SUSTAINABILITY

Specifications

Concentration 2X
Content And Storage 2X qPCR Master Mix is stored at –20°C. Nuclease-free water may be stored at any temperature.
Detection Method Primer-probe
Form Liquid
Format Tube
PCR Method qPCR
Polymerase AmpliTaq Gold DNA Polymerase
Reaction Speed Fast or Standard
Passive Reference Dye ROX (Pre-mixed)
Product Line Ambion, Path-ID
Product Type qPCR Master Mix
Purity or Quality Grade UP (Ultra Pure)
Quantity 100 reactions
Sufficient For 100 Reactions
For Use With (Application) Animal Health, Pathogen Detection
Hot Start Built-In Hot Start
No. of Reactions 100 Reactions
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.