missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Applied Biosystems™ Path-ID™ Multiplex One-Step RT-PCR Kit
Shop All Applied Biosystems Products
Click to view available options
No. of Reactions:
100 Reactions
1000 Reactions
500 Reactions
Unit Size:
100 reactions
1000 reactions
500 reactions
Description
The Path-ID™ Multiplex One-Step RT-PCR Kit is optimized for the amplification of up to four targets simultaneously using your RNA samples and TaqMan™ primer-probe sets. Reactions are run using a single-tube, one-step procedure to reverse-transcribe the RNA and amplify your targets. The kit contains a Multiplex Enzyme Mix with Arrayscript™ reverse transcriptase and AmpliTaq Gold™ DNA Polymerase, and a buffer mix.
Specifications
Specifications
| Content And Storage | Store at -20°C. Includes 100 reactions with the following components: Multiplex RT-PCR Buffer, Multiplex Enzyme Mix, and Nuclease-free water. |
| Detection Method | Primer-probe |
| Format | Tube |
| GC-Rich PCR Performance | Low |
| PCR Method | 1-step RT-qPCR, Multiplex qPCR |
| Polymerase | AmpliTaq Gold DNA Polymerase |
| Reaction Speed | Standard |
| For Use With (Equipment) | 7500 Fast System, 7500 System |
| Passive Reference Dye | ROX (Pre-mixed) |
| Product Line | Path-ID |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction