missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Patent Blue VF, pure, indicator grade

Product Code. 10457750 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10457750 25 g 25g
15274664 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10457750 Supplier Thermo Scientific Chemicals Supplier No. 189410250
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

CAS: 129-17-9 | C27H31N2NaO6S2 | 566.66 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Indicator
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 129-17-9
Molecular Formula C27H31N2NaO6S2
Molecular Weight (g/mol) 566.66
MDL Number MFCD00012119
InChI Key SJEYSFABYSGQBG-UHFFFAOYSA-M
Synonym Acid Blue 1, C.I. 42045, αzurine 2G, Sulphan blue
PubChem CID 8507
ChEBI CHEBI:34906
IUPAC Name sodium;4-[[4-(diethylamino)phenyl]-(4-diethylazaniumylidenecyclohexa-2,5-dien-1-ylidene)methyl]benzene-1,3-disulfonate
SMILES [Na+].CCN(CC)C1=CC=C(C=C1)[C+](C1=CC=C(C=C1)N(CC)CC)C1=CC=C(C=C1S([O-])(=O)=O)S([O-])(=O)=O

Specifications

Solubility Information Solubility in water: 50g/L (20°C). Other solubilities: 40g/L (95°C), 30 mg/l in egme, 7 mg/l in etoh
Infrared Spectrum Authentic
Formula Weight 566.67
Grade Pure
Packaging Glass bottle
Suitability suitable as acid-base indicator
UV Spectrometry Complies
Color Blue/Green to Purple
Quantity 25 g
Physical Form Crystalline Powder
Chemical Name or Material Patent Blue VF
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.