missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Pateclizumab Recombinant Monoclonal Antibody
Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA559854
This item is not returnable.
View return policy
Description
For reconstitution, add sterile, distilled water to achieve a final antibody concentration of 1 mg/mL. Gently shake to solubilize the protein completely. Do not vortex. Reconstituted products should be stored at -80 °.
Pateclizumab is under investigation in clinical trial NCT01225393 (A Study to Evaluate the Efficacy and Safety of MLTA3698A in Combination With a Disease-Modifying Anti-Rheumatic pharmaceutical (DMARD) Compared With Adalimumab in Combination With a DMARD in Patients With Active Rheumatoid Arthritis).
Specifications
| Pateclizumab Humanized | |
| Recombinant Monoclonal | |
| Unconjugated | |
| MLTA3698A; PRO283698; RG7416 | |
| Protein A | |
| RUO | |
| Human | |
| Antibody | |
| IgG1 |
| ELISA, Flow Cytometry, Functional Assay, Surface Plasmon Resonance | |
| 1 mg/mL | |
| 25 mM histidine with 8% sucrose, 0.01% Tween 80 and no preservative; pH 6.2 | |
| Human | |
| 1 mg | |
| Primary | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction