missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Patched Domain Containing 2 Antibody, FITC, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-81756F
This item is not returnable.
View return policy
Description
Patched Domain Containing 2 Polyclonal antibody specifically detects Patched Domain Containing 2 in Human samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| Patched Domain Containing 2 | |
| Polyclonal | |
| PBS | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| FITC | |
| DISP3, Dispatched 3, KIAA1337, Patched Domain-Containing Protein 2, Protein Dispatched Homolog 3, PTCHD2 | |
| Patched Domain Containing 2 antibody was raised against a 14 amino acid synthetic peptide from near the carboxy terminus of human Patched Domain Containing 2. The immunogen is located within amino acids 1240 - 1290 of Patched Domain Containing 2. Amino Acid Squence: LAGENLPPHQAEDA | |
| 0.1 mL | |
| Signal Transduction | |
| 57540 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction