missing translation for 'onlineSavingsMsg'
Learn More

Patched Domain Containing 2 Antibody - BSA Free, Novus Biologicals™

Product Code. 18382970 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18382970 0.1 mg 0.10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18382970 Supplier Novus Biologicals Supplier No. NBP281756

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Patched Domain Containing 2 Polyclonal specifically detects Patched Domain Containing 2 in Human samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Patched Domain Containing 2
Applications ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution ELISA, Immunohistochemistry 5 μg/mL, Immunohistochemistry-Paraffin
Formulation PBS
Gene Alias DISP3, Dispatched 3, KIAA1337, Patched Domain-Containing Protein 2, Protein Dispatched Homolog 3, PTCHD2
Gene Symbols PTCHD2
Host Species Rabbit
Immunogen Patched Domain Containing 2 antibody was raised against a 14 amino acid synthetic peptide from near the carboxy terminus of human Patched Domain Containing 2. The immunogen is located within amino acids 1240 - 1290 of Patched Domain Containing 2. Amino Acid Squence: LAGENLPPHQAEDA
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57540
Test Specificity At least two isoforms of Patched Domain Containing 2 are known to exist. This antibody is specific for Patched Domain Containing 2 and will not recognize the other DISP family of proteins.
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.