missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Patched 2/PTCH2 Antibody (OTI8B5), Alexa Fluor™ 350, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-73255AF350
This item is not returnable.
View return policy
Description
Patched 2/PTCH2 Monoclonal specifically detects Patched 2/PTCH2 in Human, Mouse samples. It is validated for Western Blot.
Specifications
| Patched 2/PTCH2 | |
| Monoclonal | |
| Alexa Fluor 350 | |
| 50 mM sodium borate with 0.05% sodium azide | |
| PTCH2 | |
| Recombinant protein fragment corresponding to amino acids 79-392 of human Patched 2/PTCH2 (NP_003729) produced in E.coli. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse | |
| IgG1 |
| Western Blot | |
| OTI8B5 | |
| Western Blot | |
| patched (Drosophila) homolog 2, patched 2, patched homolog 2, patched homolog 2 (Drosophila), protein patched homolog 2, PTC2 | |
| Mouse | |
| Affinity Purified | |
| Cancer | |
| 8643.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction