missing translation for 'onlineSavingsMsg'
Learn More

Patched 1/PTCH Antibody, mFluor Violet 610 SE, Novus Biologicals™

Product Code. 18407233
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18407233 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18407233 Supplier Novus Biologicals (Bio-Techne) Supplier No. NBP171662MFV610

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Patched 1/PTCH Polyclonal antibody specifically detects Patched 1/PTCH in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen Patched 1/PTCH
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate mFluor Violet 610 SE
Dilution Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias BCNSFLJ42602, FLJ26746, HPE7, NBCCS, patched (Drosophila) homolog, patched 1, patched homolog (Drosophila), patched homolog 1, patched homolog 1 (Drosophila), protein patched homolog 1, PTC, PTC1, PTCH, PTCH protein +12b, PTCH protein +4', PTCH protein -10, PTCH11
Host Species Rabbit
Immunogen A genomic peptide made to a C-terminal region of the human Patched 1 protein (within residues 1200-1350). [Swiss-Prot Q13635]
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Cycle and Replication, Stem Cell Signaling Pathway
Primary or Secondary Primary
Gene ID (Entrez) 5727
Target Species Human, Mouse, Rat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.