missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Pasteurella multocida Toxin (PMT) Detection Kit

Product Code. 13206069
Click to view available options
Quantity:
96 Tests (1 Plate)
960 Tests (10 Plates)
Unit Size:
96 tests
This item is not returnable. View return policy

Product Code. 13206069

Brand: Thermo Scientific™ K0009119

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Rapidly detect Pasteurella multocida toxin, the causal factor of progressive atrophic rhinitis in pigs with this assay.

Detect Pasteurella multocida toxin, the causal factor of progressive atrophic rhinitis in pigs1, with Thermo Scientific™ Oxoid™ Pasteurella multocida Toxin PMT Kit. The enzyme-linked immunosorbent assay is designed to deliver specific, sensitive, and fast detection of Pasteurella multocida toxin in porcine samples.

Progressive atrophic rhinitis is a disease of pigs, caused by infection with toxigenic P. multocida.1 The pathogenesis of the disease is linked to a single, well-characterized component: the Pasteurella multocida toxin (PMT). The clinical manifestations of the disease are shortening or distortion of the snout, sneezing, nasal discharge, and epitaxis. Reduced growth rate is seen in severe cases. Use ELISA Systems Pasteurella multocida Toxin PMT Detection Kit for the detection of Pasteurella multocida toxin in porcine samples.
  • Simple: Ready-to-use liquid reagents and concentrates make the assay easy to implement and run
  • Sensitive: Detect proteins at low levels for improved assurance of safety
  • Specific: Detect PMT in any type of sample, including extracts of primary mixed cultures obtained from, e.g., nasal swabs or washings2
Other names for the PMT include dermonecrotic toxin (DNT), heat labile toxin (HLT), turbinate atrophy toxin (TAT), and osteolytic toxin. For Veterinary Use Only. Not all products are available for sale in all territories. Please inquire. Remel™ and Oxoid™ products are now part of the Thermo Scientific™ brand.
TRUSTED_SUSTAINABILITY

Specifications

Description Pasteurella multocida Toxin (PMT) Detection Kit
Detectable Analytes Pasteurella multocida toxin
Immunoassay Kit Format ELISA (Standard)
Kit Contents Conjugate, Microtitration plate, Positive control antigen, Reconstitution buffer, Sealing tape, Stop solution, Substrate, Wash buffer concentrate, White strips
Quantity 96 Tests (1 Plate)
Storage Requirements 2°C to 8°C
compliance-icons compliance-icons
Signal Word
  • Danger
Hazard Category
  • Reproductive toxicity Category 1B
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H360D-May damage the unborn child.
  • H315-Causes skin irritation.
  • H318-Causes serious eye damage.
Precautionary Statement
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.