missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PASK Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 14396-1-AP-150UL
This item is not returnable.
View return policy
Description
PASKIN (PAS-Kinase) is thought to regulate protein synthesis in cells. It is a conserved gene product found in yeast as well as mammals. PASKIN contains two domains (PAS A and PAS B) as well as a serine/threonine kinase domain related to AMP kinases. PASKIN activity leads to decreased carbohydrate storage, as well as increased protein synthesis. PASKIN-dependent phosphorylation inhibits the activity of mammalian glycogen synthase.Specifications
| PASK | |
| Polyclonal | |
| Unconjugated | |
| PASK | |
| hPASK, KIAA0135, PAS kinase, PASK, PASKIN, STK37 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 23178, 269224, 301617 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| 0.26 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q8CEE6, Q96RG2 | |
| PASK | |
| PASK Fusion Protein Ag5893 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction