missing translation for 'onlineSavingsMsg'
Learn More

IBA Lifesciences pASK-IBA E. coli vector
GREENER_CHOICE

Product Code. p-7236938 Shop All IBA Lifesciences Products
Change view
Click to view available options
Vector:
pASK-IBA2C
pASK-IBA3C
pASK-IBA4C
pASK-IBA5C
pASK-IBA6C
pASK-IBA7C
Unit Size:
5µg
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Vector unitSize
12806486 pASK-IBA2C 5µg
12826486 pASK-IBA4C 5µg
12836486 pASK-IBA5C 5µg
12846486 pASK-IBA6C 5µg
12856486 pASK-IBA7C 5µg
12816486 pASK-IBA3C 5µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Product Code. 12806486 Supplier IBA Lifesciences Supplier No. 21321000

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

pASK-IBA E. coli expression vectors encoding a Strep-tag™II and chloramphenicol resistance

pASK-IBA vectors are classic cloning vectors devised for periplasmic or cytoplasmic E. coli expression of recombinant proteins with the Strep-tag™II fused to the C- or N-terminus. They carry the chloramphenicol resistance cassette for selection of transformed E. coli cells, F1 and ColE1 origin for a high plasmid copy number, a multiple cloning site with recognition sites for several common restriction enzymes (e.g. EcoRI, SmaI, BamHI, SalI, PstI) and the inducible tetracycline promoter/operator for the regulated expression of proteins. Some carry the ompA signal for periplasmic secretion of the recombinant protein.The vector can be combined with any E. coli strain since the tet-promoter works independently of the genetic background of E. coli. Expression of the recombinant protein is induced by addition of anhydrotetracycline, which is a derivative of tetracycline that does not exhibit antibiotic activity. During secretion into the periplasmic space the ompA signal sequence will be cleaved off.

Specifications

Format Suspension
Promoter Tet
Storage Buffer TE buffer
Antibiotic Resistance Bacterial Chloramphenicol
Expression System E. coli
Protein Tag Position (to your gene) C-terminal
Replication Origin F1 and ColE1 origin
Size Range approx. 3 kb
Special Elements ompA signal for periplasmic secretion
Protein Tag Strep-tag II
Cloning Method Direct Cloning
Concentration 250 ng/μL
Vector pASK-IBA2C
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.