missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Pascolizumab Humanized Recombinant Human Monoclonal Antibody

Human Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA541725
This item is not returnable.
View return policy
Description
Endotoxin level < 0.01EU/μg by LAL method Use a manual defrost freezer and avoid repeated freeze thaw cycles. Store at 2 to 8 °C for one week. Store at -20 to -80 °C for twelve months from the date of receipt.
Pascolizumab (SB 240683) is a humanized anti-interleukin-4 antibody with therapeutic potential in asthma. Pascolizumab is a humanized anti-IL-4 monoclonal antibody that can inhibit upstream and downstream events associated with asthma, including (T(H)2) cell activation and immunoglobulin E production.
Specifications
| Pascolizumab Humanized | |
| Recombinant Monoclonal | |
| Unconjugated | |
| SB-240683 | |
| Human IL4. | |
| 100 μg | |
| Primary | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid | |
| Chemical |
| ELISA, SDS-Page | |
| 2.15 mg/mL | |
| PBS with no preservative; pH 7.5 | |
| Human | |
| Protein A | |
| RUO | |
| Human | |
| Antibody | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction